Learn More
Medchemexpress LLC GPR120 modulator 1 | 1050506-75-6 | 98.06% | C19H16ClNO4S | 50 MG
SDP
Catalog No. 5000359211
Encompass_Preferred
GPR120 modulator 1 is a G protein-coupled receptor 120 (GPR120) modulator, extracted from patent US8394841B2, compound example F1. It is intended for research use only and can be used for the study of diseases associated with abnormal or deregulated GPR120, such as diabetes.
  • G protein-coupled receptor 120 (GPR120) modulator
  • Extracted from patent US8394841B2, compound example F1
  • Useful for research of diseases associated with abnormal or deregulated GPR120, such as diabetes
  • For research use only
Catalog No. 50-003-59211 Supplier Medchemexpress LLC Supplier No. HY5016250MG
GPR120 modulator 1 is a G protein-coupled receptor 120 (GPR120) modulator, extracted from patent US8394841B2, compound example F1. It is intended for research use only and can be used for the study of diseases associated with abnormal or deregulated GPR120, such as diabetes.
  • G protein-coupled receptor 120 (GPR120) modulator
  • Extracted from patent US8394841B2, compound example F1
  • Useful for research of diseases associated with abnormal or deregulated GPR120, such as diabetes
  • For research use only