Learn More
Medchemexpress LLC GPR120 modulator 1 | 1050506-75-6 | 98.1% | 389.85 g/mol | C19H16ClNO4S | 10 MG
SDP
Catalog No. 5000298611
Encompass_Preferred
GPR120 modulator 1 is a small-molecule research compound used to modulate G protein-coupled receptor 120 (GPR120) in studies of metabolic and inflammatory pathways. It is intended for in vitro and preclinical research use and is supplied as a purified solid with reported purity suitable for biochemical assays.
  • Small-molecule modulator of GPR120 for receptor biology studies.
  • High reported purity for reliable assay performance.
  • Available in small pack sizes suitable for screening and characterization.
  • Provided with supporting documents including datasheet and safety information.
Catalog No. 50-002-98611 Supplier Medchemexpress LLC Supplier No. HY5016210MG
GPR120 modulator 1 is a small-molecule research compound used to modulate G protein-coupled receptor 120 (GPR120) in studies of metabolic and inflammatory pathways. It is intended for in vitro and preclinical research use and is supplied as a purified solid with reported purity suitable for biochemical assays.
  • Small-molecule modulator of GPR120 for receptor biology studies.
  • High reported purity for reliable assay performance.
  • Available in small pack sizes suitable for screening and characterization.
  • Provided with supporting documents including datasheet and safety information.