Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GPR120 modulator 2 | 1050506-87-0 | 97.1% | 387.88 | C20H18ClNO3S | 10 MG
SDP

Catalog No. 5000298466
Encompass_Preferred

GPR120 modulator 2 is a small-molecule research compound that modulates the free fatty acid receptor GPR120 and is intended for in vitro pharmacology and metabolic signaling studies.

  • Targets the free fatty acid receptor GPR120.
  • Molecular formula C20H18ClNO3S and molecular weight 387.88.
  • CAS number 1050506-87-0 for unambiguous identification.
  • High purity (97.1%) suitable for research applications.
  • Available in small pack sizes, including 10 mg.
  • Storage: powder at -20°C for long-term stability, 4°C short term; in solvent store at -80°C.

Catalog No. 50-002-98466 Supplier Medchemexpress LLC Supplier No. HY5017210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPR120 modulator 2 is a small-molecule research compound that modulates the free fatty acid receptor GPR120 and is intended for in vitro pharmacology and metabolic signaling studies.

  • Targets the free fatty acid receptor GPR120.
  • Molecular formula C20H18ClNO3S and molecular weight 387.88.
  • CAS number 1050506-87-0 for unambiguous identification.
  • High purity (97.1%) suitable for research applications.
  • Available in small pack sizes, including 10 mg.
  • Storage: powder at -20°C for long-term stability, 4°C short term; in solvent store at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.