Learn More
Aviva Systems Biology Corp GPR137 Antibody - N-terminal region (ARP49476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR137. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human GPR137. Alternate names: C11orf4, GPR137A, TM7SF1L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-429