Learn More
Aviva Systems Biology Corp GPR161 Antibody - N-terminal region (ARP42353_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42353_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR161. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GPR161. Alternate names: FLJ33952, RE2ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58194