Learn More
Aviva Systems Biology Corp GPR161 Antibody - middle region (ARP42355_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42355_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR161. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPR161. Alternate names: FLJ33952, RE2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58196