Learn More
Aviva Systems Biology Corp GPR177 Antibody - middle region (ARP44432_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44432_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR177. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPR177. Alternate names: C1orf139, EVI, FLJ23091, MGC131760, MGC14878, MRP, WLS, GPR177ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53423