Learn More
Aviva Systems Biology Corp GPR177 Antibody - N-terminal region (ARP44430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR177. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GPR177. Alternate names: RP11-518D3.2, C1orf139, EVI, FLJ23091, MGC131760, MGC14878, MRP, WLS, GPR177ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53422