Learn More
TARGETMOL CHEMICALS INC GPR183 5MG
Supplier: TARGETMOL CHEMICALS INC T99755MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GPR183 (3-(34-difluorophenyl)-N-[3-fluoro-5-(morpholin-4-yl)phenyl]propanamide) is a chemotactic receptor known for its role in the maturation of B cells and the endogenous ligand is the oxysterol 7(alpha)25-dihydroxycholesterol (7(alpha)25-OHC). GPR183 can be used in research on inflammatory bowel disease (IBD). purity: 98%