Learn More
Aviva Systems Biology Corp GPR22 Antibody - C-terminal region (ARP65761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR22. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human GPR22. Alternate names: MGC129847ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-564