Learn More
Aviva Systems Biology Corp GPR27 Antibody - middle region (ARP47954_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47954_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR27. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GPR27. Alternate names: SREB1ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-285-334