Learn More
Medchemexpress LLC GPR30 agonist-1 | 415919-74-3 | 98.6% | 374.25 | 50 MG
SDP
Catalog No. 5000349106
Encompass_Preferred
GPR30 agonist-1 is a G protein-coupled receptor 30 (GPR30) agonist that exerts vasorelaxant effects.
  • Induces a concentration-dependent relaxation in carotid arteries from both male and female rats.
  • Relaxation is abolished by endothelium removal.
  • Relaxation is abrogated in the presence of the nitric oxide synthase inhibitor NG-nitro-l-arginine methyl ester (100 μM).
Catalog No. 50-003-49106 Supplier Medchemexpress LLC Supplier No. HY13868650MG
GPR30 agonist-1 is a G protein-coupled receptor 30 (GPR30) agonist that exerts vasorelaxant effects.
  • Induces a concentration-dependent relaxation in carotid arteries from both male and female rats.
  • Relaxation is abolished by endothelium removal.
  • Relaxation is abrogated in the presence of the nitric oxide synthase inhibitor NG-nitro-l-arginine methyl ester (100 μM).