Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GPR30 agonist-1 | 415919-74-3 | MFCD02316399 | 98.6% | 374.25 g/mol | C19H17BrFNO | 5 MG
SDP

Catalog No. 5000221099
Encompass_Preferred

GPR30 agonist-1 is a small-molecule agonist of G protein-coupled receptor 30 (GPR30/GPER) used in biochemical and pharmacological research. It has reported vasorelaxant activity and is supplied as a purified powder for in vitro and in vivo studies. The compound is soluble in DMSO and is provided in multiple vial sizes for research use.

  • Selective agonist of GPR30, suitable for receptor signaling studies.
  • Reported to exert vasorelaxant effects in vascular tissue.
  • High purity (≈98.6%) suitable for research applications.
  • Molecular formula C19H17BrFNO; molecular weight 374.25 g/mol.
  • Soluble in DMSO at 50 mg/mL with ultrasonic warming.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-21099 Supplier Medchemexpress LLC Supplier No. HY1386865MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPR30 agonist-1 is a small-molecule agonist of G protein-coupled receptor 30 (GPR30/GPER) used in biochemical and pharmacological research. It has reported vasorelaxant activity and is supplied as a purified powder for in vitro and in vivo studies. The compound is soluble in DMSO and is provided in multiple vial sizes for research use.

  • Selective agonist of GPR30, suitable for receptor signaling studies.
  • Reported to exert vasorelaxant effects in vascular tissue.
  • High purity (≈98.6%) suitable for research applications.
  • Molecular formula C19H17BrFNO; molecular weight 374.25 g/mol.
  • Soluble in DMSO at 50 mg/mL with ultrasonic warming.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.