Learn More
Eurofins Discoverx PathHunter GPR35 Activated GPCR Internalization U2OS Cell Line, 2 vials. (93-0968C3)
Catalog No. 502001708
Encompass
The PathHunter GPR35 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of GPR35 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged GPR35. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting GPR35-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1708 Supplier Eurofins Discoverx Supplier No. 930968C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter GPR35 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of GPR35 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged GPR35. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting GPR35-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.