Learn More
TARGETMOL CHEMICALS INC GPR35 AGONIST 2 10MG
SDP
Catalog No. 502580871
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GPR35 agonist 2 (TC-G 1001) is a potent GPR35 agonist.The EC50 values of GPR35 agonist 2 in the (beta)-arrestin and Ca2+ release assays were 26 and 3.2 nM respectively. purity: 98%
Catalog No. 50-258-0871 Supplier TARGETMOL CHEMICALS INC Supplier No. T2343410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GPR35 agonist 2 (TC-G 1001) is a potent GPR35 agonist.The EC50 values of GPR35 agonist 2 in the (beta)-arrestin and Ca2+ release assays were 26 and 3.2 nM respectively. purity: 98%