Learn More
Aviva Systems Biology Corp GPR35 Antibody - C-terminal region (ARP64657_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64657_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR35. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human GPR35. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-288-326