Learn More
Medchemexpress LLC HY-103083 5mg Medchemexpress, GPR40 agonist 4 CAS:2102196-57-4 Purity:>98%
SDP
Catalog No. 501968029
Encompass_Preferred
Medchemexpress, HY-103083 5mg GPR40 agonist 4 CAS:2102196-57-4 GPR40 agonist 4 is a potent free fatty acid receptor 1 (FFA1/ GPR40) agonist with a pEC50 of 7.54. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8029 Supplier Medchemexpress LLC Supplier No. HY1030835MG
Medchemexpress, HY-103083 5mg GPR40 agonist 4 CAS:2102196-57-4 GPR40 agonist 4 is a potent free fatty acid receptor 1 (FFA1/ GPR40) agonist with a pEC50 of 7.54. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.