Learn More
Eurofins Discoverx ChemiBrite FFA1/GPR40 Free Fatty Acid Receptor Stable Cell Line, 2 vials. (HTS038L)
Catalog No. 502005722
Encompass
The ChemiBrite FFA1/GPR40 Free Fatty Acid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of FFAR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FFAR1, a naturally Gq coupled GPCR. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of FFAR1, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5722 Supplier Eurofins Discoverx Supplier No. HTS038L
Add to Cart
Add to Cart
Request a Quote
The ChemiBrite FFA1/GPR40 Free Fatty Acid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of FFAR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FFAR1, a naturally Gq coupled GPCR. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of FFAR1, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.