Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GPR40/FFAR1 modulator 1 | 874755-26-7 | 98.28% | 389.41 | 50 MG
SDP

Catalog No. 5000437086
Encompass_Preferred

GPR40/FFAR1 modulator 1 acts as an agonist and an allosteric modulator for Gq-coupled free fatty acid receptor 1 (GPR40/FFAR1). This product is intended for research use only.

  • Agonist and allosteric modulator for Gq-coupled free fatty acid receptor 1.
  • Purity of 98.28%.
  • Solid appearance.
  • Off-white to light yellow color.
  • Soluble in DMSO (31.25 mg/mL).

Catalog No. 50-004-37086 Supplier Medchemexpress LLC Supplier No. HY11176350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPR40/FFAR1 modulator 1 acts as an agonist and an allosteric modulator for Gq-coupled free fatty acid receptor 1 (GPR40/FFAR1). This product is intended for research use only.

  • Agonist and allosteric modulator for Gq-coupled free fatty acid receptor 1.
  • Purity of 98.28%.
  • Solid appearance.
  • Off-white to light yellow color.
  • Soluble in DMSO (31.25 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.