Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GPR40/FFAR1 modulator 1 | 874755-26-7 | 98.3% | 389.41 | 10 MG
SDP

Catalog No. 5000438891
Encompass_Preferred

GPR40/FFAR1 modulator 1 is an agonist and an allosteric modulator for Gq-coupled free fatty acid receptor 1 (GPR40/FFAR1).

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protected from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-004-38891 Supplier Medchemexpress LLC Supplier No. HY11176310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

GPR40/FFAR1 modulator 1 is an agonist and an allosteric modulator for Gq-coupled free fatty acid receptor 1 (GPR40/FFAR1).

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protected from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.