Learn More
Aviva Systems Biology Corp GPR52 Antibody - C-terminal region (ARP62835_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62835_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR52. It was validated on Western Blot. . Alternate names: MGC111751ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-287-781