Learn More
Cayman Chemical GPR52 Comp-43, Purity: ≥98%, CAS: 1239987-91-7, 50 mg
SDP
Catalog No. 502947348
Encompass_Preferred
A GPR52 antagonist (IC50 = 0.63 µM); reduces mHTT protein levels in STHdhQ7/Q111 cells in a concentration-dependent manner; reduces apoptosis induced by growth factor deprivation in primary striatal neurons isolated from HdhQ7/Q140 mice; decreases striatal levels of soluble and insoluble mHTT, as well as increases the latency to fall in the rotarod test, in the HdhQ140 mouse model of Huntington's disease at 5 mg/kg
Catalog No. 50-294-7348 Supplier Cayman Chemical Supplier No. 3356450 MG
A GPR52 antagonist (IC50 = 0.63 µM); reduces mHTT protein levels in STHdhQ7/Q111 cells in a concentration-dependent manner; reduces apoptosis induced by growth factor deprivation in primary striatal neurons isolated from HdhQ7/Q140 mice; decreases striatal levels of soluble and insoluble mHTT, as well as increases the latency to fall in the rotarod test, in the HdhQ140 mouse model of Huntington's disease at 5 mg/kg