Learn More
Eurofins Discoverx Ready-to-Assay GPR54/KiSS1 Receptor Frozen Cells, 2 Vials of Cells. (HTS032RTA)
Catalog No. 502005708
Encompass
The Ready-to-Assay GPR54/KiSS1 Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand based activation of KISS1R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses KISS1R alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of KISS1R, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5708 Supplier Eurofins Discoverx Supplier No. HTS032RTA
The Ready-to-Assay GPR54/KiSS1 Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand based activation of KISS1R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses KISS1R alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of KISS1R, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.