Learn More
Aviva Systems Biology Corp ADGRG1 Antibody - N-terminal region (ARP58627_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58627_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR56. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GPR56. Alternate names: BFPP, DKFZp781L1398, TM7LN4, TM7XN1ApplicationWB. Species reactivity: Human, Rabbit, Rat
Catalog No. 50-014-62381