Learn More
Aviva Systems Biology Corp GPR6 Antibody - N-terminal region (ARP42229_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42229_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GPR6. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-014-58153