Learn More
Proteintech Group Inc GPR65 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 203061AP150UL
Encompass_Preferred
The GPR65 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR65. This antibody recognizes human antigen. The GPR65 antibody has been validated for the following applications: IHC, ELISA analysis.
Catalog No. 50-225-6429