Learn More
Aviva Systems Biology Corp GPR68 Antibody - middle region (ARP64436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR68. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human GPR68. Alternate names: MGC111379, MGC156983, OGR1, GPR12AApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-262