Learn More
Aviva Systems Biology Corp GPR75 Antibody - middle region (ARP33104_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33104_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR75. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPR75. Alternate names: GPR-chr2, WI-31133, GPRchr2, WI31133ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54887