Learn More
Santa Cruz Biotechnology GPR75 siRNA and shRNA Plasmids (h) 10 µM
Catalog No. NC3881602
Encompass
Target species: human; for corresponding mouse product, see GPR75 siRNA and shRNA Plasmids (m)Gene Silencers generally consist of pools of three to five target-specific 19-25 nucleotide sequences in lengthFor independent verification of GPR75 gene silencing results, individual duplex components or plasmids are also available upon requestGet details about si/shRNA Gene SilencersGene silencers are intended for noncommercial and research use only.
Catalog No. NC3881602 Supplier Santa Cruz Biotechnology Supplier No. SC94341
Target species: human; for corresponding mouse product, see GPR75 siRNA and shRNA Plasmids (m)Gene Silencers generally consist of pools of three to five target-specific 19-25 nucleotide sequences in lengthFor independent verification of GPR75 gene silencing results, individual duplex components or plasmids are also available upon requestGet details about si/shRNA Gene SilencersGene silencers are intended for noncommercial and research use only.