Learn More
Aviva Systems Biology Corp GPR83 Antibody - C-terminal region (ARP59122_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59122_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR83. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GPR83. Alternate names: GIR, GPR72ApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-286-587