Learn More
Aviva Systems Biology Corp GPR87 Antibody - N-terminal region (ARP42620_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42620_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR87. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GPR87. Alternate names: FKSG78, GPR95, KPG_002, MGC131898ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-62941