Learn More
Aviva Systems Biology Corp GPR87 Antibody - middle region (ARP42621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR87. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPR87. Alternate names: FKSG78, GPR95, KPG_002, MGC131898ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62942