Learn More
Aviva Systems Biology Corp Anti-GPR88 Antibody 100 µl
SDP
Supplier:  Aviva Systems Biology Corp ARP42607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gpr88. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Gpr88ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-011