Learn More
Aviva Systems Biology Corp GPRASP2 Antibody - middle region (ARP52604_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52604_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPRASP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPRASP2. Alternate names: FLJ35662, FLJ37327, GASP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60781