Learn More
OriGene GPRC5D (NM_018654) Human Over-expression Lysate, 20 ug
SDP
Supplier:  OriGene LC412961
Encompass_Preferred
GPRC5D HEK293T cell transient overexpression lysate (as WB positive control)
Catalog No. NC2873908