Learn More
Aviva Systems Biology Corp GPRC6A Antibody - N-terminal region (ARP64455_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64455_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPRC6A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human GPRC6A. Alternate names: GPCR, bA86F4.3ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-266