Learn More
Carnabio Usa Inc GPRK6(GRK6)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 013155
Encompass
GPRK6(GRK6), Full-length, Wild type, Amino Acid 1-560, NP_001004105.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0449