Learn More
Carnabio Usa Inc GPRK7(GRK7)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0111610
Encompass
GPRK7(GRK7), Full-length, Wild type, Amino Acid 1-553, NP_631948.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0806