Learn More
Enzo Life Sciences G protein alphai3 subunit polyclonal antibody (50 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA1290050
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 345-354 of human Gαi3 subunit. Species reactivity: Human Bovine. Specificity: Recognizes the αi3 subunit of G protein. Applications: IP, WB. Purity: Protein A affinity purified IgG. Formulation: Liquid. In 100mM potassium phosphate, pH 7.5, containing 140mM sodium chloride. Contains no preservatives.
Catalog No. 50-201-1759