Learn More
Medchemexpress LLC GPRP acetate | 157009-81-9 | 98.9% | 485.53 | 25 MG
SDP
Catalog No. 5000425832
Encompass_Preferred
GPRP acetate (Gly-Pro-Arg-Pro acetate) is a fibrin polymerization inhibitor that blocks the interaction of fibrinogen with the platelet membrane glycoprotein IIb/IIIa complex (GPIIb/IIIa). This compound enhances free thrombin generation and has demonstrated significant therapeutic potential in animal models, alleviating DSS-induced colitis by improving body weight loss and mortality.
  • Inhibits fibrin polymerization.
  • Targets platelet membrane glycoprotein IIb/IIIa complex.
  • Increases free thrombin generation.
  • Alleviates DSS-induced colitis in animal models.
  • Reduces DSS-induced shortening of colon length.
  • Decreases tissue damage and infiltration of inflammatory cells.
  • Lowers levels of inflammatory cytokines in colitis mice.
Catalog No. 50-004-25832 Supplier Medchemexpress LLC Supplier No. 25MGHYP0074A
GPRP acetate (Gly-Pro-Arg-Pro acetate) is a fibrin polymerization inhibitor that blocks the interaction of fibrinogen with the platelet membrane glycoprotein IIb/IIIa complex (GPIIb/IIIa). This compound enhances free thrombin generation and has demonstrated significant therapeutic potential in animal models, alleviating DSS-induced colitis by improving body weight loss and mortality.
  • Inhibits fibrin polymerization.
  • Targets platelet membrane glycoprotein IIb/IIIa complex.
  • Increases free thrombin generation.
  • Alleviates DSS-induced colitis in animal models.
  • Reduces DSS-induced shortening of colon length.
  • Decreases tissue damage and infiltration of inflammatory cells.
  • Lowers levels of inflammatory cytokines in colitis mice.