Learn More
Medchemexpress LLC HY-P0074A 5mg , GPRP (acetate) CAS:157009-81-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0074A5MG
Encompass_Preferred
HY-P0074A 5mg GPRP (acetate) CAS: 157009-81-9 GPRP acetate (Gly-Pro-Arg-Pro acetate) is a fibrin polymerization inhibitor that inhibits the interaction of fibrinogen with the platelet membrane glycoprotein IIb/IIIa complex (GPIIb/IIIa). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1561