Learn More
TARGETMOL CHEMICALS INC GPS491 5MG
SDP
Catalog No. 502579996
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GPS491 has antiviral activity and inhibits HIV-1 adenovirus and coronavirus replication by altering RNA processing. purity: 98%
Catalog No. 50-257-9996 Supplier TARGETMOL CHEMICALS INC Supplier No. T402935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GPS491 has antiviral activity and inhibits HIV-1 adenovirus and coronavirus replication by altering RNA processing. purity: 98%