Learn More
Aviva Systems Biology Corp GPSN2 Antibody - middle region (ARP48431_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48431_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPSN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPSN2. Alternate names: SC2, TER, GPSN2, MRT14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53552