Learn More
Aviva Systems Biology Corp GPT Antibody - N-terminal region (ARP45712_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45712_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GPT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GPT. Alternate names: AAT1, ALT1, GPT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59114