Learn More
Medchemexpress LLC GpTx-1 TFA | 99.6% | 4073.82 | 500 UG
SDP
Catalog No. 5000424359
Encompass_Preferred
GpTx-1 TFA is a peptide-based NaV1.7 sodium channel antagonist isolated from the venom of the Chilean spider Grammostola porter. It demonstrates potent inhibitory activity against the NaV1.7 channel with an IC50 value of 10 nM.
  • Shows excellent selectivity for NaV1.4 and NaV1.5.
  • Exhibits greater than 20-fold selectivity for NaV1.4.
  • Exhibits greater than 950-fold selectivity for NaV1.5.
Catalog No. 50-004-24359 Supplier Medchemexpress LLC Supplier No. HYP1681A500UG
GpTx-1 TFA is a peptide-based NaV1.7 sodium channel antagonist isolated from the venom of the Chilean spider Grammostola porter. It demonstrates potent inhibitory activity against the NaV1.7 channel with an IC50 value of 10 nM.
  • Shows excellent selectivity for NaV1.4 and NaV1.5.
  • Exhibits greater than 20-fold selectivity for NaV1.4.
  • Exhibits greater than 950-fold selectivity for NaV1.5.