Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Gradient Valve Standard, 5 mL, Propyl Paraben, Methanol Solvent

Catalog No. 50818872 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit is a Gradient Valve Standard designed for the assessment of the Alliance Gradient Proportioning Valve.

  • Kit contains a 1x5 mL standard solution.
  • Active ingredient: propyl paraben at 0.56 g/L (±5%) with ≥99% purity.
  • Solvent: 100% HPLC grade methanol.
  • Separation mode: reversed phase; detection method: UV.
  • Formulation: liquid; storage condition: room temperature.
  • System type: LC; quantitative analysis is supported.
  • Shelf life: 36 months; shipping condition: ambient.
  • Volume: 5 mL; type: reference standard; application: system performance.
  • Units per package: 1.

Catalog No. 50-818-872 Supplier Waters Corp Supplier No. WAT042876
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit is a Gradient Valve Standard designed for the assessment of the Alliance Gradient Proportioning Valve.

  • Kit contains a 1x5 mL standard solution.
  • Active ingredient: propyl paraben at 0.56 g/L (±5%) with ≥99% purity.
  • Solvent: 100% HPLC grade methanol.
  • Separation mode: reversed phase; detection method: UV.
  • Formulation: liquid; storage condition: room temperature.
  • System type: LC; quantitative analysis is supported.
  • Shelf life: 36 months; shipping condition: ambient.
  • Volume: 5 mL; type: reference standard; application: system performance.
  • Units per package: 1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.