Learn More
Waters Corp Tubing Kit, GPV, For Alliance Separations Modules
Catalog No. 50793063 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation GPV Tubing Kit is designed for use with Alliance 2690/2695 and 2790/2795 Separations Modules. It provides replacement tubing for the gradient proportioning valve (GPV) outlet.
  • Used to replace GPV outlet tubing in Alliance separations modules.
  • Enables customization for low- or high-flow applications.
  • Suitable for replacing clogged standard tubing.
  • Helps provide appropriate volume for mixing needs.
Catalog No. 50-793-063 Supplier Waters Corp Supplier No. 700004868
The Waters Corporation GPV Tubing Kit is designed for use with Alliance 2690/2695 and 2790/2795 Separations Modules. It provides replacement tubing for the gradient proportioning valve (GPV) outlet.
  • Used to replace GPV outlet tubing in Alliance separations modules.
  • Enables customization for low- or high-flow applications.
  • Suitable for replacing clogged standard tubing.
  • Helps provide appropriate volume for mixing needs.