Learn More
AdipoGen GPX1 human pAb
SDP
Supplier:  AdipoGen AG25A0104C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human GPX1. Detects a band of ~23kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. GPX1 is an antioxidant enzyme that is down-regulated in cancer cells. Protects the hemoglobin in erythrocytes from oxidative breakdown. Its main role is the prevention of cancer initiation by ROS-mediated DNA damage.
Catalog No. 50-256-3216