Learn More
AdipoGen GPX1 human rec His
SDP
Catalog No. 502562972
Encompass_Preferred
Protein. Human GPX1 aa 1-201 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. In PBS. Purity >90% SDS-PAGE. Glutathione peroxidase 1 GPX1 is an antioxidant enzyme that is down-regulated in cancer cells. Protects the hemoglobin in erythrocytes from oxidative breakdown. Its main role is the prevention of cancer initiation by ROS-mediated DNA damage.
Catalog No. 50-256-2972 Supplier AdipoGen Supplier No. AG40A0150Y3010
Protein. Human GPX1 aa 1-201 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. In PBS. Purity >90% SDS-PAGE. Glutathione peroxidase 1 GPX1 is an antioxidant enzyme that is down-regulated in cancer cells. Protects the hemoglobin in erythrocytes from oxidative breakdown. Its main role is the prevention of cancer initiation by ROS-mediated DNA damage.