Learn More
Aviva Systems Biology Corp GPX2 Antibody - middle region (ARP42018_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GPX2. Alternate names: GI-GPx, GPRP, GSHPX-GI, GSHPx-2, GPx-2, GPRP-2, GPx-GIApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-284-972